Ethyl-5-aminopyrazine-2-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 5-aminopyrazine-2-carboxylate
H66918-03
H66918-MD
AAH66918-03EA
409.22
CAD
AAH66918-03
AAH66918-MD
Ethyl-5-aminopyrazine-2-carboxylate 95%
Ethyl-5-aminopyrazine-2-carboxylate
Purity: 95%
CAS No.: 54013-06-8
Molecular Formula: C7H9N3O2
Formula Weight: 167.17
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 609566
MDL No.: MFCD08437665
Formula:
C₇H₉N₃O₂ MW: 167.17 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08437665 CAS Number: 54013-06-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...