BAPTA tetramethyl ester 99%
Supplier: Thermo Scientific Chemicals
H66907-03
H66907-06
H66907-MD
AAH66907-03EA
131.14
CAD
AAH66907-03
AAH66907-06
AAH66907-MD
BAPTA tetramethyl ester 99%
BAPTA tetramethyl ester
Purity: 99%
CAS No.: 125367-34-2
Molecular Formula: C26H32N2O10
Formula Weight: 532.55
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00532664
Formula:
C₂₆H₃₂N₂O₁₀ Storage Temperature: Ambient |
MDL Number:
MFCD00532664 CAS Number: 125367-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...