N-Tosyl-l-proline 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tos-Pro-OH, (2S)-1-[(4-methylphenyl)sulphonyl]pyrrolidine-2-carboxylic acid, N-Toluenesulfonyl-L-proline, N-Toluenesulphonyl-L-proline, (2S)-1-[(4-methylphenyl)sulfonyl]pyrrolidine-2-carboxylic acid

H66902-06 H66902-14
AAH66902-06EA 111.86 CAD
AAH66902-06 AAH66902-14
N-Tosyl-l-proline 94%
N-Tosyl-l-proline

Purity: 95% CAS No.: 51077-01-1 Molecular Formula: C12H15NO4S Formula Weight: 269.32 Storage Temperature: -30°C to -10°C Physical Form: Low melting solid UN No.: 2811 MDL No.: MFCD00238392

Formula: C₁₂H₁₅NO₄S
MW: 269.315 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238392
CAS Number: 51077-01-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR