5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H66885-03 H66885-06 H66885-MD
AAH66885-03EA 104.28 CAD
AAH66885-03 AAH66885-06 AAH66885-MD
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 76041-79-7 Molecular Formula: C6H3BrF3NO Formula Weight: 242.00 Physical Form: Solid

Formula: C₆H₃BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD12546486
CAS Number: 76041-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR