2-Bromonicotinonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H66881-03 H66881-06
AAH66881-03EA 52.61 CAD
AAH66881-03 AAH66881-06
2-Bromonicotinonitrile 98%
2-Bromonicotinonitrile

Purity: 98% CAS No.: 20577-26-8 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Physical Form: Solid

Formula: C₆H₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD00234140
CAS Number: 20577-26-8
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR