2-Bromo-5-methylpyrimidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H66877-03 H66877-MD
AAH66877-03EA 557.74 CAD
AAH66877-03 AAH66877-MD
2-Bromo-5-methylpyrimidine 95%
2-Bromo-5-methylpyrimidine

Purity: 95% CAS No.: 150010-20-1 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Physical Form: Solid

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753737
CAS Number: 150010-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR