2-Hydroxy-4-nitropyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Nitropyridin-2-ol
H66852-03
H66852-MD
AAH66852-03EA
274.92
CAD
AAH66852-03
AAH66852-MD
2-Hydroxy-4-nitropyridine 96%
2-Hydroxy-4-nitropyridine
Purity: 96%
CAS No.: 4487-51-8
Molecular Formula: C5H4N2O3
Formula Weight: 140.10
Flash Point: 156°(313°F)
Physical Form: Liquid
Density: 1.44
Appearance: Colorless to yellow
Boiling Point: 334°
MDL No.: MFCD04114149
Formula:
C₅H₄N₂O₃ Boiling Pt: 334 °C Density: 1.44 Flash Pt: 156 °C(313 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04114149 CAS Number: 4487-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...