(2-Oxo-3-benzoxazolyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(2-Oxobenzo[d]oxazol-3(2H)-yl)acetic acid
H66828-03
H66828-06
AAH66828-03EA
95.43
CAD
AAH66828-03
AAH66828-06
(2-Oxo-3-benzoxazolyl)acetic acid 97%
(2-Oxo-3-benzoxazolyl)acetic acid
Purity: 97%
CAS No.: 13610-49-6
Molecular Formula: C9H7NO4
Formula Weight: 193.16
Physical Form: Solid
Formula:
C₉H₇NO₄ MW: 193.158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01547452 CAS Number: 13610-49-6 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...