4-Bromo-2,6-dichlorophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

H66763-03 H66763-06
AAH66763-03EA 40.13 CAD
AAH66763-03 AAH66763-06
4-Bromo-2,6-dichlorophenol 97%
4-Bromo-2,6-dichlorophenol

Purity: 97% CAS No.: 3217-15-0 Molecular Formula: C6H3BrCl2O Formula Weight: 241.89 Physical Form: Powder Appearance: Off-white Beilstein Reference: 2046857 UN No.: 2811 MDL No.: MFCD00019996

Formula: C₆H₃BrCl₂O
MW: 241.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019996
CAS Number: 3217-15-0
EINECS: 221-740-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR