1-Benzhydrylazetidin-3-ol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H66720-14
H66720-22
AAH66720-14EA
73.31
CAD
AAH66720-14
AAH66720-22
1-Benzhydrylazetidin-3-ol 97%
1-Benzhydrylazetidin-3-ol
Purity: 97%
CAS No.: 18621-17-5
Molecular Formula: C16H17NO
Formula Weight: 239.32
Physical Form: Solid
Formula:
C₁₆H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD00205109 CAS Number: 18621-17-5 UN: 3259 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...