Methyl-5-bromopyrazine-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 5-bromopyrazine-2-carboxylate

H66706-03 H66706-MD
AAH66706-03EA 334.96 CAD
AAH66706-03 AAH66706-MD
Methyl-5-bromopyrazine-2-carboxylate 97%
Methyl 5-bromopyrazine-2-carboxylate

Purity: 97% CAS No.: 210037-58-4 Molecular Formula: C6H5BrN2O2 Formula Weight: 217.02 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD09991715

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09991715
CAS Number: 210037-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR