6-Bromo[1,2,4]triazolo[1,5-a]pyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-[1,2,4]triazolo[1,5-a]pyridine

H66700-03 H66700-MD
AAH66700-03EA 178.54 CAD
AAH66700-03 AAH66700-MD
6-Bromo[1,2,4]triazolo[1,5-a]pyridine 96%
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine

Purity: 96% CAS No.: 356560-80-0 Molecular Formula: C6H4BrN3 Formula Weight: 198.02 Physical Form: Solid

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689534
CAS Number: 356560-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR