Methyl-6-chloropyrazine-2-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 6-chloropyrazine-2-carboxylate
H66682-03
H66682-MD
AAH66682-03EA
233.84
CAD
AAH66682-03
AAH66682-MD
Methyl-6-chloropyrazine-2-carboxylate 95%
Methyl-6-chloropyrazine-2-carboxylate
Purity: 95%
CAS No.: 23611-75-8
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 880043
MDL No.: MFCD10686598
Formula:
C₆H₅ClN₂O₂ Storage Temperature: Freezer |
MDL Number:
MFCD10686598 CAS Number: 23611-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...