1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
H66672-03
H66672-MD
AAH66672-03EA
467.68
CAD
AAH66672-03
AAH66672-MD
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester
Purity: 97%
CAS No.: 1151802-22-0
Molecular Formula: C12H21BN2O3
Formula Weight: 234.11
Physical Form: Solid
Formula:
C₁₂H₁₉BN₂O₂ MW: 234.106 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16659007 CAS Number: 1151802-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...