Ethyl benzimidazole-4-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

H66669-03 H66669-MD
AAH66669-03EA 437.66 CAD
AAH66669-03 AAH66669-MD
Ethyl benzimidazole-4-carboxylate 95%
Ethyl benzimidazole-4-carboxylate

Purity: 95% CAS No.: 167487-83-4 Molecular Formula: C10H10N2O2 Formula Weight: 190.20 Physical Form: Solid

Formula: C₁₀H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18251681
CAS Number: 167487-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR