2-(2-Methyl-6-nitrophenyl)acetic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66654-03
H66654-MD
AAH66654-03EA
222.78
CAD
AAH66654-03
AAH66654-MD
2-(2-Methyl-6-nitrophenyl)acetic acid 95%
2-(2-Methyl-6-nitrophenyl)acetic acid
Purity: 95%
CAS No.: 23876-18-8
Molecular Formula: C9H9NO4
Formula Weight: 195.17
Physical Form: Solid
Formula:
C₉H₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00066313 CAS Number: 23876-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...