1,2,3,4-Tetrahydronaphthalen-2-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

H66652-03 H66652-06
AAH66652-03EA 328.64 CAD
AAH66652-03 AAH66652-06
1,2,3,4-Tetrahydronaphthalen-2-amine 97%
1,2,3,4-Tetrahydronaphthalen-2-amine

Purity: 97% CAS No.: 2954-50-9 Molecular Formula: C10H13N Formula Weight: 147.1 Physical Form: Solid

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045577
CAS Number: 2954-50-9
EINECS: 220-973-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR