2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Nitisinone, 2-(2-Nitro-4-(trifluoromethyl)benzoyl)cyclohexane-1,3-dione

H66633-03 H66633-MD
AAH66633-03EA 491.38 CAD
AAH66633-03 AAH66633-MD
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione 95%
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione

Purity: 95% CAS No.: 104206-65-7 Molecular Formula: C14H10F3NO5 Formula Weight: 329.23 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD01752192

Formula: C₁₄H₁₀F₃NO₅
MW: 329.231 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01752192
CAS Number: 104206-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR