Ethyl-5-chloropyrazine-2-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 5-chloropyrazine-2-carboxylate

H66630-03 H66630-MD
AAH66630-03EA 257.54 CAD
AAH66630-03 AAH66630-MD
Ethyl-5-chloropyrazine-2-carboxylate 95%
Ethyl-5-chloropyrazine-2-carboxylate

Purity: 95% CAS No.: 54013-04-6 Molecular Formula: C7H7ClN2O2 Formula Weight: 186.60 Physical Form: Solid

Formula: C₇H₇ClN₂O₂
MW: 186.595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18070850
CAS Number: 54013-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR