2-Amino-6-fluorophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66626-03
H66626-06
AAH66626-03EA
73.79
CAD
AAH66626-03
AAH66626-06
AAH66626-MD
2-Amino-6-fluorophenol 97%
2-Amino-6-fluorophenol
Purity: 97%
CAS No.: 53981-25-2
Molecular Formula: C6H6FNO
Formula Weight: 127.12
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 3603077
UN No.: 2811
MDL No.: MFCD08532465
Formula:
C₆H₆FNO Storage Temperature: Freezer |
MDL Number:
MFCD08532465 CAS Number: 53981-25-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...