5-Methyl-2-(2,2,2-trimethylacetamido)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(5-Methylpyridin-2-yl)pivalamide
H66596-03
H66596-06
H66596-MD
AAH66596-03EA
104.28
CAD
AAH66596-03
AAH66596-06
AAH66596-MD
5-Methyl-2-(2,2,2-trimethylacetamido)pyridine 98%
5-Methyl-2-(2,2,2-trimethylacetamido)pyridine
Purity: 98%
CAS No.: 86847-78-1
Molecular Formula: C11H16N2O
Formula Weight: 192.26
Physical Form: Solid
Formula:
C₁₁H₁₆N₂O MW: 192.262 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03374646 CAS Number: 86847-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...