3-Bromotetrahydro-4H-pyran-4-one 90%

Supplier: Thermo Scientific Chemicals
Warning

H66595-03 H66595-MD
AAH66595-03EA 467.68 CAD
AAH66595-03 AAH66595-MD
3-Bromotetrahydro-4H-pyran-4-one 90%
3-Bromotetrahydro-4H-pyran-4-one

Purity: 90% CAS No.: 98021-79-5 Molecular Formula: C5H7BrO2 Formula Weight: 179.01 Physical Form: Solid

Formula: C₅H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD13193743
CAS Number: 98021-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR