tert-Butyl (3-oxocyclohexyl)carbamate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Boc-amino)cyclohexanone
H66594-03
H66594-06
H66594-MD
AAH66594-03EA
221.2
CAD
AAH66594-03
AAH66594-06
AAH66594-MD
tert-Butyl (3-oxocyclohexyl)carbamate 95%
tert-Butyl (3-oxocyclohexyl)carbamate
Purity: 95%
CAS No.: 885280-38-6
Molecular Formula: C11H19NO3
Formula Weight: 213.28
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 11040732
MDL No.: MFCD03790905
Formula:
C₁₁H₁₉NO₃ MW: 213.27 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03790905 CAS Number: 885280-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...