6-Aminomethylquinoline 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-(Aminomethyl)quinoline

H66548-03 H66548-MD
AAH66548-03EA 541.94 CAD
AAH66548-03 AAH66548-MD
6-Aminomethylquinoline 95%
6-Aminomethylquinoline

Purity: 95% CAS No.: 99071-54-2 Molecular Formula: C10H10N2 Formula Weight: 158.20 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 115231 MDL No.: MFCD02853688

Formula: C₁₀H₁₀N₂
MW: 158.204 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02853688
CAS Number: 99071-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR