1-(2,3-Epoxypropyl)-2-nitroimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66536-03
H66536-MD
AAH66536-03EA
715.74
CAD
AAH66536-03
AAH66536-MD
1-(2,3-Epoxypropyl)-2-nitroimidazole 97%
1-(2,3-Epoxypropyl)-2-nitroimidazole
Purity: 97%
CAS No.: 13551-90-1
Molecular Formula: C6H7N3O3
Formula Weight: 169.14
Physical Form: Solid
Formula:
C₆H₇N₃O₃ MW: 169.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20527194 CAS Number: 13551-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...