4-Chloro-3-(hydroxymethyl)pyridine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Chloropyridin-3-yl)methanol hydrochloride
H66522-03
H66522-MD
AAH66522-03EA
451.88
CAD
AAH66522-03
AAH66522-MD
4-Chloro-3-(hydroxymethyl)pyridine hydrochloride 98%
4-Chloro-3-(hydroxymethyl)pyridine hydrochloride
Purity: 98%
CAS No.: 245124-17-8
Molecular Formula: C6H6ClNO.HCl
Formula Weight: 180.03
Physical Form: Solid
Formula:
C₆H₇Cl₂NO MW: 180.028 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD23703142 CAS Number: 245124-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...