3-Chloro-5-methylaniline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66518-03
H66518-06
H66518-MD
AAH66518-03EA
79.63
CAD
AAH66518-03
AAH66518-06
AAH66518-MD
3-Chloro-5-methylaniline 95%
3-Chloro-5-methylaniline
Purity: 95%
CAS No.: 29027-20-1
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 742113
UN No.: 2239
MDL No.: MFCD15527197
Formula:
C7H8ClN Storage Temperature: Freezer |
MDL Number:
MFCD15527197 CAS Number: 29027-20-1 UN: 3429 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...