4-Hydroxy-6-methylnicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66499-03
H66499-06
AAH66499-03EA
331.8
CAD
AAH66499-03
AAH66499-06
4-Hydroxy-6-methylnicotinic acid 97%
4-Hydroxy-6-methylnicotinic acid
Purity: 97%
CAS No.: 33821-58-8
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Physical Form: Solid
Formula:
C₇H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD04038153 CAS Number: 33821-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...