Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, white ≥97.0%, solid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine
H66485-03
H66485-MD
AAH66485-03EA
578.28
CAD
AAH66485-03
AAH66485-MD
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, white ≥97.0%, solid
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Purity: 97+%
CAS No.: 510758-28-8
Molecular Formula: C30H30N10
Formula Weight: 530.63
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 132-143°
MDL No.: MFCD09265124
Formula:
C₃₀H₃₀N₁₀ MW: 530.64 g/mol Melting Pt: 132…143 °C Storage Temperature: Freezer |
MDL Number:
MFCD09265124 CAS Number: 510758-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...