6-Chloro-2-fluoronicotinic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66484-03
H66484-MD
AAH66484-03EA
633.58
CAD
AAH66484-03
AAH66484-MD
6-Chloro-2-fluoronicotinic acid 95%
6-Chloro-2-fluoronicotinic acid
Purity: 95%
CAS No.: 1211578-46-9
Molecular Formula: C6H3ClFNO2
Formula Weight: 175.54
Physical Form: Solid
Formula:
C₆H₃ClFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD16610397 CAS Number: 1211578-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...