2-Amino-2-(5-indolyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-2-(1H-indol-5-yl)acetic acid

H66482-03 H66482-MD
AAH66482-03EA 383.94 CAD
AAH66482-03 AAH66482-MD
2-Amino-2-(5-indolyl)acetic acid 98%
2-Amino-2-(5-indolyl)acetic acid

Purity: 98% CAS No.: 108763-43-5 Molecular Formula: C10H10N2O2 Formula Weight: 190.20 Physical Form: Powder Appearance: Pink Beilstein Reference: 15714312 MDL No.: MFCD06656882

Formula: C₁₀H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06656882
CAS Number: 108763-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR