5-Bromo-2-methylpyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66467-03
H66467-MD
AAH66467-03EA
161.16
CAD
AAH66467-03
AAH66467-MD
5-Bromo-2-methylpyrimidine 98%
5-Bromo-2-methylpyrimidine
Purity: 98%
CAS No.: 7752-78-5
Molecular Formula: C5H5BrN2
Formula Weight: 173.01
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 108736á
MDL No.: MFCD07375143
Formula:
C₅H₅BrN₂ MW: 173.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07375143 CAS Number: 7752-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...