1-Methylbenzoimidazol-5-amine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66465-03
H66465-06
AAH66465-03EA
262.28
CAD
AAH66465-03
AAH66465-06
1-Methylbenzoimidazol-5-amine 97%
1-Methylbenzoimidazol-5-amine
Purity: 97%
CAS No.: 10394-38-4
Molecular Formula: C8H9N3
Formula Weight: 147.08
Physical Form: Solid
Formula:
C₈H₉N₃ Storage Temperature: Ambient |
MDL Number:
MFCD03164349 CAS Number: 10394-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...