5-Aminophthalide 97%

Supplier: Thermo Scientific Chemicals
Warning

H66457-03 H66457-06
AAH66457-03EA 351.39 CAD
AAH66457-03 AAH66457-06
5-Aminophthalide 97%
5-Aminophthalide

Purity: 97% CAS No.: 65399-05-5 Molecular Formula: C8H7NO2 Formula Weight: 149.05 Physical Form: Solid

Formula: C₈H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00778315
CAS Number: 65399-05-5
EINECS: 265-731-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR