2-Bromonaphthalen-1-ol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-1-naphthalenol
H66429-03
H66429-06
H66429-MD
AAH66429-03EA
143.46
CAD
AAH66429-03
AAH66429-06
AAH66429-MD
2-Bromonaphthalen-1-ol 95%
2-Bromonaphthalen-1-ol
Purity: 95%
CAS No.: 771-15-3
Molecular Formula: C10H7BrO
Formula Weight: 223.07
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 2554660
MDL No.: MFCD00274158
Formula:
C₁₀H₇BrO MW: 223.07 g/mol Melting Pt: 44…46 °C Storage Temperature: Freezer |
MDL Number:
MFCD00274158 CAS Number: 771-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...