2'-Hydroxy-4'-methylacetanilide 97%

Supplier: Thermo Scientific Chemicals
Warning

H66423-03 H66423-MD
AAH66423-03EA 114.87 CAD
AAH66423-03 AAH66423-MD
2'-Hydroxy-4'-methylacetanilide 97%
2'-Hydroxy-4'-methylacetanilide

Purity: 97% CAS No.: 13429-10-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Solid

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00012061
CAS Number: 13429-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR