5-Bromo-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H66411-03 H66411-MD
AAH66411-03EA 404.48 CAD
AAH66411-03 AAH66411-MD
5-Bromo-1H-pyrazolo[3,4-b]pyridine 95%
5-Bromo-1H-pyrazolo[3,4-b]pyridine

Purity: 95% CAS No.: 875781-17-2 Molecular Formula: C6H4BrN3 Formula Weight: 198.02 Physical Form: Solid

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663982
CAS Number: 875781-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR