3-Methylindole-2-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methyl-1H-indole-2-carbaldehyde
H66393-03
H66393-MD
AAH66393-03EA
413.96
CAD
AAH66393-03
AAH66393-MD
3-Methylindole-2-carboxaldehyde 97%
3-Methylindole-2-carboxaldehyde
Purity: 97%
CAS No.: 5257-24-9
Molecular Formula: C10H9NO
Formula Weight: 159.18
Physical Form: Solid
Sensitivity: Air and Moisture Sensitive
Beilstein Reference: 4034
MDL No.: MFCD00957066
Formula:
C₁₀H₉NO MW: 159.188 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00957066 CAS Number: 5257-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...