5-Bromo-2-pyrazinecarboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromopyrazine-2-carboxylic acid
H66382-03
H66382-MD
AAH66382-03EA
499.28
CAD
AAH66382-03
AAH66382-MD
5-Bromo-2-pyrazinecarboxylic acid 95%
5-Bromo-2-pyrazinecarboxylic acid
Purity: 95%
CAS No.: 876161-05-6
Molecular Formula: C5H3BrN2O2
Formula Weight: 203.00
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD09909672
Formula:
C₅H₃BrN₂O₂ Storage Temperature: Freezer |
MDL Number:
MFCD09909672 CAS Number: 876161-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...