1-(2-Thienyl)piperidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66330-03
H66330-06
H66330-MD
AAH66330-03EA
94.48
CAD
AAH66330-03
AAH66330-06
AAH66330-MD
1-(2-Thienyl)piperidine 98%
1-(2-Thienyl)piperidine
Purity: 95%
CAS No.: 19983-20-1
Molecular Formula: C9H13NS
Formula Weight: 167.27
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Orange
Beilstein Reference: 1239029
MDL No.: MFCD00142961
Formula:
C₉H₁₃NS Storage Temperature: Ambient |
MDL Number:
MFCD00142961 CAS Number: 19983-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...