3-Amino-6-cyano-2-methylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-6-methylpicolinonitrile

H66325-03 H66325-MD
AAH66325-03EA 483.48 CAD
AAH66325-03 AAH66325-MD
3-Amino-6-cyano-2-methylpyridine 95%
3-Amino-6-cyano-2-methylpyridine

Purity: 95% CAS No.: 1079054-78-6 Molecular Formula: C7H7N3 Formula Weight: 133.15 Physical Form: Solid

Formula: C₇H₇N₃
MW: 133.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062828
CAS Number: 1079054-78-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR