Methyl-5-chloropyrazine-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 5-chloropyrazine-2-carboxylate

H66306-03 H66306-06
AAH66306-03EA 46.77 CAD
AAH66306-03 AAH66306-06
Methyl-5-chloropyrazine-2-carboxylate 98%
Methyl 5-chloropyrazine-2-carboxylate

Purity: 98% CAS No.: 33332-25-1 Molecular Formula: C6H5ClN2O2 Formula Weight: 172.57 Physical Form: Solid

Formula: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01632102
CAS Number: 33332-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR