3-Amino-2-methoxypyridine-4-carboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66289-03
H66289-MD
AAH66289-03EA
526.14
CAD
AAH66289-03
AAH66289-MD
3-Amino-2-methoxypyridine-4-carboxylic acid 95%
3-Amino-2-methoxypyridine-4-carboxylic acid
Purity: 95%
CAS No.: 870997-81-2
Molecular Formula: C7H8N2O3
Formula Weight: 168.15
Physical Form: Solid
Formula:
C₇H₈N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD18262266 CAS Number: 870997-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...