3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66286-03
H66286-06
AAH66286-03EA
34.44
CAD
AAH66286-03
AAH66286-06
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 76041-71-9
Molecular Formula: C6H3ClF3NO
Formula Weight: 197.54
Physical Form: Solid
Melting Point: 159-161°
Beilstein Reference: 5932693
MDL No.: MFCD00153095
Formula:
C₆H₃ClF₃NO Melting Pt: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153095 CAS Number: 76041-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...