2-(2-Cyanophenoxy)acetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

H66271-03 H66271-06 H66271-14
AAH66271-03EA 38.71 CAD
AAH66271-03 AAH66271-06 AAH66271-14
2-(2-Cyanophenoxy)acetamide 98%
2-(2-Cyanophenoxy)acetamide

Purity: 98% CAS No.: 54802-12-9 Molecular Formula: C9H8N2O2 Formula Weight: 176.17 Physical Form: Solid

Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00297091
CAS Number: 54802-12-9
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR