2-Oxo-1-pyrrolidineacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Oxo-1-pyrrolidinyl)acetic acid
H66260-03
H66260-06
AAH66260-03EA
254.38
CAD
AAH66260-03
AAH66260-06
2-Oxo-1-pyrrolidineacetic acid 97%
2-Oxo-1-pyrrolidineacetic acid
Purity: 97%
CAS No.: 53934-76-2
Molecular Formula: C6H9NO3
Formula Weight: 143.14
Physical Form: Solid
Formula:
C₆H₉NO₃ MW: 143.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00962836 CAS Number: 53934-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...