1-(cyclopropylcarbonyl)piperazine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
H66242-03
H66242-06
H66242-MD
AAH66242-03EA
99.38
CAD
AAH66242-03
AAH66242-06
AAH66242-MD
1-(cyclopropylcarbonyl)piperazine hydrochloride 97%
1-(cyclopropylcarbonyl)piperazine hydrochloride
Purity: 97%
CAS No.: 1021298-67-8
Molecular Formula: C8H14N2O.HCl
Formula Weight: 190.67
Physical Form: Solid
Appearance: White
Melting Point: 175-179°
Beilstein Reference: 18312533
MDL No.: MFCD03428576
Formula:
C₈H₁₅ClN₂O Melting Pt: 175…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD03428576 CAS Number: 1021298-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...