4-Bromo-1-trityl-1H-Pyrazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66225-03
H66225-06
AAH66225-03EA
62.25
CAD
AAH66225-03
AAH66225-06
4-Bromo-1-trityl-1H-Pyrazole 95%
4-Bromo-1-trityl-1H-Pyrazole
Purity: 95%
CAS No.: 95162-14-4
Molecular Formula: C22H17BrN2
Formula Weight: 389.30
Physical Form: Powder
Appearance: White
Beilstein Reference: 550183
MDL No.: MFCD09907863
Formula:
C₂₂H₁₇BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09907863 CAS Number: 95162-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...