3-(Trifluoroacetyl)pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanol
H66214-03
H66214-MD
AAH66214-03EA
458.2
CAD
AAH66214-03
AAH66214-MD
3-(Trifluoroacetyl)pyridine 95%
3-(Trifluoroacetyl)pyridine
Purity: 95%
CAS No.: 138624-99-4
Molecular Formula: C7H4F3NO
Formula Weight: 175.11
Physical Form: Solid
Formula:
C₇H₆F₃NO MW: 177.126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16067922 CAS Number: 138624-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...