6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione 98%

Supplier: Thermo Scientific Chemicals
Warning

H66212-03 H66212-06
AAH66212-03EA 62.25 CAD
AAH66212-03 AAH66212-06
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione 98%
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione

Purity: 98% CAS No.: 194421-56-2 Molecular Formula: C7H9NO2 Formula Weight: 139.15 Physical Form: Solid

Formula: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12923551
CAS Number: 194421-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR